COMH0C955833 Combi-Blocks, Inc.

Benzyldimethylstearylammonium chloride hydrate

  • MDL number MFCD00149986
  • Cas Number : 122-19-0
  • Molecular Weight : 442.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7663

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-7663
Availability details for
Quantity
  • Availability
  • Add to Cart