COMH0C95586D Combi-Blocks, Inc.

1-Chloro-4-[chloro(4-chlorophenyl)methyl]benzene

  • MDL number MFCD00972762
  • Cas Number : 782-08-1
  • Molecular Weight : 271.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6849

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6849
Availability details for
Quantity
  • Availability
  • Add to Cart