COMH0C955876 Combi-Blocks, Inc.

2-(Piperidin-1-ylcarbonyl)benzoic acid

  • MDL number MFCD00443957
  • Cas Number : 20320-44-9
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5572

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5572
Availability details for
Quantity
  • Availability
  • Add to Cart