COMH11E51CCC Combi-Blocks, Inc.

6-Bromo-3-chloro-2-methylpyridine-4-boronic acid

  • MDL number MFCD31653711
  • Cas Number : 2377606-09-0
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1290

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1290
Availability details for
Quantity
  • Availability
  • Add to Cart