COMH11E51CCF Combi-Blocks, Inc.

2,3-Difluoro-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number MFCD31653750
  • Cas Number : 2345641-33-8
  • Molecular Weight : 265.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1450

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1450
Availability details for
Quantity
  • Availability
  • Add to Cart