COMH11E51CD1 Combi-Blocks, Inc.

tert-Butyl N-[2-(3-methoxyphenoxy)ethyl]-N-methylcarbamate

  • MDL number MFCD31653784
  • Cas Number : 2237237-78-2
  • Molecular Weight : 281.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2121

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2121
Availability details for
Quantity
  • Availability
  • Add to Cart