COMH11E51CD4 Combi-Blocks, Inc.

3-(Dimethylamino)-4-methanesulfonylbenzoic acid

  • MDL number MFCD31653792
  • Cas Number : 2237235-55-9
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2164

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2164
Availability details for
Quantity
  • Availability
  • Add to Cart