COMH11E51CD5 Combi-Blocks, Inc.

N-ethyl-2-fluoro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

  • MDL number MFCD31653839
  • Molecular Weight : 293.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2863

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2863
Availability details for
Quantity
  • Availability
  • Add to Cart