COMH11E51CD7 Combi-Blocks, Inc.

1-{[2-fluoro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbonyl}pyrrolidine

  • MDL number MFCD31653841
  • Molecular Weight : 319.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2874

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2874
Availability details for
Quantity
  • Availability
  • Add to Cart