COMH11E51CD9 Combi-Blocks, Inc.

2-fluoro-N-phenyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

  • MDL number MFCD31653843
  • Molecular Weight : 341.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2876

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2876
Availability details for
Quantity
  • Availability
  • Add to Cart