COMH11E51CDF Combi-Blocks, Inc.

5-Bromo-4-chloro-2-fluoro-3-methylphenylboronic acid

  • MDL number MFCD31653900
  • Cas Number : 2377606-52-3
  • Molecular Weight : 267.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6917

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6917
Availability details for
Quantity
  • Availability
  • Add to Cart