COMH11E51CE9 Combi-Blocks, Inc.

2-[2-(dimethylamino)ethoxy]-5-fluorobenzonitrile

  • MDL number MFCD18599940
  • Cas Number : 1405023-08-6
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5839

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5839
Availability details for
Quantity
  • Availability
  • Add to Cart