COMH11E51CEC Combi-Blocks, Inc.

Methyl 4-fluoro-5-methyl-2-nitrobenzoate

  • MDL number MFCD28797657
  • Cas Number : 1437780-12-5
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6025

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6025
Availability details for
Quantity
  • Availability
  • Add to Cart