COMH11E51CF3 Combi-Blocks, Inc.

4-Cyano-2,6-difluorophenylboronic acid

  • MDL number MFCD03095140
  • Cas Number : 2199440-52-1
  • Molecular Weight : 182.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1058

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1058
Availability details for
Quantity
  • Availability
  • Add to Cart