COMH11E51CF4 Combi-Blocks, Inc.

4-Cyano-2,3-difluorophenylboronic acid

  • MDL number MFCD07644471
  • Cas Number : 2377608-51-8
  • Molecular Weight : 182.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1450

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1450
Availability details for
Quantity
  • Availability
  • Add to Cart