COMH11E51CF5 Combi-Blocks, Inc.

N-BOC-1H-Isoindoline-5-boronic acid

  • MDL number MFCD14581766
  • Cas Number : 2152653-47-7
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2978

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2978
Availability details for
Quantity
  • Availability
  • Add to Cart