COMH11E51CF7 Combi-Blocks, Inc.

5-Methylthiophene-3-boronic acid

  • MDL number MFCD18250237
  • Cas Number : 930303-82-5
  • Molecular Weight : 141.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-1485

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-1485
Availability details for
Quantity
  • Availability
  • Add to Cart