COMH11E51CF9 Combi-Blocks, Inc.

Methyl 3-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate

  • MDL number MFCD16996413
  • Cas Number : 2377610-23-4
  • Molecular Weight : 297.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7090
Availability details for
Quantity
  • Availability
  • Add to Cart