COMH11E51CFA Combi-Blocks, Inc.

4-Bromo-1-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-indole

  • MDL number MFCD16995792
  • Cas Number : 2247798-73-6
  • Molecular Weight : 336.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-8447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-8447
Availability details for
Quantity
  • Availability
  • Add to Cart