COMH11E51CFB Combi-Blocks, Inc.

2-(3,4-Dihydro-2H-1,5-benzodioxepin-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD31653749
  • Cas Number : 2246880-24-8
  • Molecular Weight : 276.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1360

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1360
Availability details for
Quantity
  • Availability
  • Add to Cart