COMH11E51CFE Combi-Blocks, Inc.

[5-fluoro-2-(piperidin-1-yl)pyridin-4-yl]boronic acid

  • MDL number MFCD28054646
  • Cas Number : 2225176-46-3
  • Molecular Weight : 224.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4464

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4464
Availability details for
Quantity
  • Availability
  • Add to Cart