COMH11E51D05 Combi-Blocks, Inc.

3-(2-Fluorophenyl)-2H-pyrazole

  • MDL number MFCD09860572
  • Cas Number : 149739-32-2
  • Molecular Weight : 162.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6819

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6819
Availability details for
Quantity
  • Availability
  • Add to Cart