COMH11E51D06 Combi-Blocks, Inc.

2,3-Dibromo-5H,6H,7H-pyrrolo[1,2-a]imidazole

  • MDL number MFCD31653755
  • Cas Number : 1781573-30-5
  • Molecular Weight : 265.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2346

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2346
Availability details for
Quantity
  • Availability
  • Add to Cart