COMH11E51D14 Combi-Blocks, Inc.

4-methyl-2-nitrobenzamide

  • MDL number MFCD18399646
  • Cas Number : 65078-05-9
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1368

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1368
Availability details for
Quantity
  • Availability
  • Add to Cart