COMH11E51D15 Combi-Blocks, Inc.

4-Fluoro-3-methyl-5-nitrobenzoic acid

  • MDL number MFCD18293742
  • Cas Number : 1341764-57-5
  • Molecular Weight : 199.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1488

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1488
Availability details for
Quantity
  • Availability
  • Add to Cart