COMH11E51D19 Combi-Blocks, Inc.

tert-Butyl N-[2-(2-bromophenoxy)ethyl]carbamate

  • MDL number MFCD18382015
  • Cas Number : 1204333-53-8
  • Molecular Weight : 316.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2113

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2113
Availability details for
Quantity
  • Availability
  • Add to Cart