COMH11E51D1E Combi-Blocks, Inc.

1-(2-Aminopropoxy)-3-bromobenzene

  • MDL number MFCD10034446
  • Cas Number : 1018586-11-2
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2123

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2123
Availability details for
Quantity
  • Availability
  • Add to Cart