COMH11E51D22 Combi-Blocks, Inc.

tert-Butyl N-[1-(4-bromophenoxy)propan-2-yl]carbamate

  • MDL number MFCD31653787
  • Cas Number : 2237237-81-7
  • Molecular Weight : 330.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2127

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2127
Availability details for
Quantity
  • Availability
  • Add to Cart