COMH11E51D25 Combi-Blocks, Inc.

2-[2-(Methylamino)ethoxy]benzonitrile

  • MDL number MFCD11199931
  • Cas Number : 737745-17-4
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2154

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2154
Availability details for
Quantity
  • Availability
  • Add to Cart