COMH11E51D28 Combi-Blocks, Inc.

2,4-dimethoxy-5-methylpyridine

  • MDL number MFCD16610897
  • Cas Number : 1227574-94-8
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2176

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2176
Availability details for
Quantity
  • Availability
  • Add to Cart