COMH11E51D2A Combi-Blocks, Inc.

Methyl 4-chloro-5-fluoropyridine-2-carboxylate

  • MDL number MFCD19443469
  • Cas Number : 1378464-20-0
  • Molecular Weight : 189.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2178

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2178
Availability details for
Quantity
  • Availability
  • Add to Cart