COMH11E51D49 Combi-Blocks, Inc.

1-Bromo-3-butoxybenzene

  • MDL number MFCD11127631
  • Cas Number : 165800-57-7
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1158

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1158
Availability details for
Quantity
  • Availability
  • Add to Cart