COMH11E51D50 Combi-Blocks, Inc.

1,3-Diethyl 2-(oxiran-2-ylmethyl)propanedioate

  • MDL number MFCD25958128
  • Cas Number : 13353-23-6
  • Molecular Weight : 216.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4500

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4500
Availability details for
Quantity
  • Availability
  • Add to Cart