COMH11E51D91 Combi-Blocks, Inc.

4,4,5,5-Tetramethyl-2-(3-methyl-4-nitrophenyl)-1,3,2-dioxaborolane

  • MDL number MFCD22576357
  • Cas Number : 590418-04-5
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1541

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1541
Availability details for
Quantity
  • Availability
  • Add to Cart