COMH11E51D95 Combi-Blocks, Inc.

2-Fluoro-N-methyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

  • MDL number MFCD18730368
  • Cas Number : 2222333-26-6
  • Molecular Weight : 279.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2862
Availability details for
Quantity
  • Availability
  • Add to Cart