COMH11E51DB7 Combi-Blocks, Inc.

4-Benzyloxypropiophenone

  • MDL number MFCD00009311
  • Cas Number : 4495-66-3
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4050

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4050
Availability details for
Quantity
  • Availability
  • Add to Cart