COMH11E51DBA Combi-Blocks, Inc.

3,6-Dichloro-2-fluorobenzonitrile

  • MDL number MFCD09025385
  • Cas Number : 916420-65-0
  • Molecular Weight : 190.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4601

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4601
Availability details for
Quantity
  • Availability
  • Add to Cart