COMH11E51DCF Combi-Blocks, Inc.

N-(3,4-Dimethoxyphenyl)-4-fluorobenzylamine

  • MDL number MFCD11924932
  • Cas Number : 1019542-69-8
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3376

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3376
Availability details for
Quantity
  • Availability
  • Add to Cart