COMH11E51DD9 Combi-Blocks, Inc.

Octylamine hydrochloride

  • MDL number MFCD00042019
  • Cas Number : 142-95-0
  • Molecular Weight : 165.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-2561
Availability details for
Quantity
  • Availability
  • Add to Cart