COMH11E51DED Combi-Blocks, Inc.

5-(2-Pyridyl)thiophene-2-sulfonyl chloride

  • MDL number MFCD00052284
  • Cas Number : 151858-64-9
  • Molecular Weight : 259.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0655

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-0655
Availability details for
Quantity
  • Availability
  • Add to Cart