COMH11E51DF3 Combi-Blocks, Inc.

2-Chloro-5-methoxy-4-nitro-benzoic acid methyl ester

  • MDL number MFCD28400734
  • Cas Number : 1349744-88-2
  • Molecular Weight : 245.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0410

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0410
Availability details for
Quantity
  • Availability
  • Add to Cart