COMH11E51DF9 Combi-Blocks, Inc.

N-Methyl-n-(2-thien-2-ylbenzyl)amine

  • MDL number MFCD07368538
  • Cas Number : 852180-66-6
  • Molecular Weight : 203.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7317

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-7317
Availability details for
Quantity
  • Availability
  • Add to Cart