COMH11E51DFD Combi-Blocks, Inc.

4-Chloro-2-(chloromethyl)-1-nitrobenzene

  • MDL number MFCD12025225
  • Cas Number : 57750-81-9
  • Molecular Weight : 206.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-6440

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-6440
Availability details for
Quantity
  • Availability
  • Add to Cart