COMH11E51DFE Combi-Blocks, Inc.

Methyl 9-methylfluorene-9-carboxylate

  • MDL number MFCD00577363
  • Cas Number : 3300-16-1
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4531

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4531
Availability details for
Quantity
  • Availability
  • Add to Cart