COMH11E51DFF Combi-Blocks, Inc.

1-(Triphenylmethyl)imidazole-4-carbonitrile

  • MDL number MFCD30474777
  • Cas Number : 340179-96-6
  • Molecular Weight : 335.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4606

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4606
Availability details for
Quantity
  • Availability
  • Add to Cart