COMH11E51E00 Combi-Blocks, Inc.

Ethyl (2E)-4,4,4-trifluoro-2-(hydroxyimino)-3-oxobutanoate

  • MDL number MFCD00092812
  • Cas Number : 94609-23-1
  • Molecular Weight : 213.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2519

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2519
Availability details for
Quantity
  • Availability
  • Add to Cart