COMH11E51E03 Combi-Blocks, Inc.

5-[(4-Methoxyphenyl)methoxy]pyridine-3-boronic acid

  • MDL number MFCD19098270
  • Cas Number : 1899036-19-1
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5413

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5413
Availability details for
Quantity
  • Availability
  • Add to Cart