COMH11E51E05 Combi-Blocks, Inc.

Methyl 2-(2-methylpropoxy)benzoate

  • MDL number MFCD27947257
  • Cas Number : 21018-09-7
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4654

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4654
Availability details for
Quantity
  • Availability
  • Add to Cart