COMH11E51E07 Combi-Blocks, Inc.

6-Ethoxypyridine-3-carbonitrile

  • MDL number MFCD09932786
  • Cas Number : 106853-78-5
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4658

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4658
Availability details for
Quantity
  • Availability
  • Add to Cart