COMH11E51E09 Combi-Blocks, Inc.

(3,6-Dihydro-2h-pyran-4-yl)-boronic acid

  • MDL number MFCD02179502
  • Cas Number : 1002127-60-7
  • Molecular Weight : 127.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5953

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5953
Availability details for
Quantity
  • Availability
  • Add to Cart